3',5'-Dimethylacetophenone ≥97.0% (by GC)
Supplier: TCI America
Synonyms:
1-(3,5-Dimethylphenyl)ethanone, 1-(3,5-Dimethylphenyl)ethan-1-one
D4868-1G
D4868-5G
TCD4868-1GEA
37
USD
TCD4868-1G
TCD4868-5G
3',5'-Dimethylacetophenone ≥97.0% (by GC)
3',5'-Dimethylacetophenone
Formula:
C₁₀H₁₂O MW: 148.2 g/mol Boiling Pt: 237°C |
MDL Number:
MFCD01075693 CAS Number: 5379-16-8 |
Specification Test Results
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 85.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...