1,3-Difluoro-5-iodobenzene ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 3,5-Difluoroiodobenzene

D4620-25G D4620-5G
TCD4620-25GEA 204 USD
TCD4620-25G TCD4620-5G
1,3-Difluoro-5-iodobenzene ≥98.0% (by GC)
1,3-Difluoro-5-iodobenzene
Formula: C₆H₃F₂I
MW: 239.99 g/mol
Boiling Pt: 60°C
Flash Pt: 82°C
MDL Number: MFCD03094155
CAS Number: 2265-91-0

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR