(Methyl)triphenylphosphonium chloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Danger

Synonyms: Methyltriphenylphosphonium chloride, Methyl-TPP-Chloride, MTPP-Chloride

M2702-25G M2702-5G
TCM2702-25GEA 230.2 USD
TCM2702-25G TCM2702-5G
(Methyl)triphenylphosphonium chloride ≥98.0% (by HPLC, titration analysis)
(Methyl)triphenylphosphonium chloride
Formula: C₁₉H₁₈ClP
MW: 312.78 g/mol
MDL Number: MFCD00797851
CAS Number: 1031-15-8
EINECS: 418-400-2
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 221.0 to 225.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR