(4-Tolyloxy)acetic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

M2729-25G M2729-5G
TCM2729-25GEA 309 USD
TCM2729-25G TCM2729-5G
(4-Tolyloxy)acetic acid ≥98.0% (by GC, titration analysis)
(4-Tolyloxy)acetic acid
Formula: (C₆H₄(CH₃))OCH₂COOH
MDL Number: MFCD00014365
CAS Number: 940-64-7
EINECS: 213-374-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 138.0 to 142.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR