1-(2,4-Difluorophenyl)-2-thiourea ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Danger

D4861-1G D4861-5G
TCD4861-1GEA 29 USD
TCD4861-1G TCD4861-5G
1-(2,4-Difluorophenyl)-2-thiourea ≥98.0% (by HPLC, total nitrogen)
1-(2,4-Difluorophenyl)-2-thiourea
Formula: (C₆H₃F₂)NC(S)NH₂
MDL Number: MFCD00041150
CAS Number: 175277-76-6
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 166.0 to 170.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR