Dansyl amide ≥98.0% (by titrimetric analysis) for albumin binding assay

Supplier: TCI America

Synonyms: Dansylamide, 5-(Dimethylamino)-1-naphthalenesulphonamide, 5-(Dimethylamino)naphthalene-1-sulphonamide

D5405-100MG D5405-25MG
TCD5405-100MGEA 44 USD
TCD5405-100MG TCD5405-25MG
Dansyl amide ≥98.0% (by titrimetric analysis) for albumin binding assay
Dansyl amide
Formula: C₁₂H₁₄N₂O₂S
MW: 250.32 g/mol
MDL Number: MFCD00004000
CAS Number: 1431-39-6
EINECS: 215-854-1

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 218.0 to 222.0 deg-C
Human serum albmin (HSA) binding activity S/N ratio min. 4
Competitive avtivity in competitive drug Remaining fluorescein inensity max. 70 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR