Mitotane ≥98.0% (by GC)

Supplier: TCI America
Danger

Synonyms: 1-Chloro-2-(2,2-dichloro-1-(4-chlorophenyl)ethyl)benzene, o,p'-DDD, 2,4'-TDE, 1-(2-Chlorophenyl)-1-(4-chlorophenyl)-2,2-dichloroethane, o,p'-TDE, 2,4'-DDD, 1-Chloro-2-[2,2-dichloro-1-(4-chlorophenyl)ethyl]benzene

M3304-1G M3304-5G
TCM3304-1GEA 98 USD
TCM3304-1G TCM3304-5G
Mitotane ≥98.0% (by GC)
Mitotane
Formula: C₁₄H₁₀Cl₄
MW: 320.04 g/mol
MDL Number: MFCD00000850
CAS Number: 53-19-0
Merck Index: 14,06216

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 76.0 to 80.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR