4-Chloro-2-(trifluoromethoxy)aniline ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 4-Chloro-2-(trifluoromethoxy)phenylamine

C3374-1G C3374-200MG
TCC3374-1GEA 106 USD
TCC3374-1G TCC3374-200MG
4-Chloro-2-(trifluoromethoxy)aniline ≥98.0% (by GC)
4-Chloro-2-(trifluoromethoxy)aniline
Formula: C₇H₅ClF₃NO
MDL Number: MFCD00153102
CAS Number: 175205-77-3

Order Now

Specification Test Results

Appearance White to Light yellow powder to lump
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR