2-Chloro-4-(trifluoromethoxy)aniline ≥97.0% (by GC, titration analysis)

Supplier: TCI America
Warning

C3142-1G C3142-5G
TCC3142-1GEA 46 USD
TCC3142-1G TCC3142-5G
2-Chloro-4-(trifluoromethoxy)aniline ≥97.0% (by GC, titration analysis)
2-Chloro-4-(trifluoromethoxy)aniline
Formula: C₇H₅ClF₃NO
MW: 211.57
Flash Pt: 98°C
MDL Number: MFCD01631704
CAS Number: 69695-61-0

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR