4-Bromo-3-nitroaniline ≥98.0% (by GC)

Supplier: TCI America
Warning

B4876-1G B4876-5G
TCB4876-1GEA 27 USD
TCB4876-1G TCB4876-5G
4-Bromo-3-nitroaniline ≥98.0% (by GC)
4-Bromo-3-nitroaniline
Formula: C₆H₅BrN₂O₂
MDL Number: MFCD01320678
CAS Number: 53324-38-2

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 126.0 to 131.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR