5-Bromoisophthalic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

B4232-1G B4232-5G
TCB4232-1GEA 42 USD
TCB4232-1G TCB4232-5G
5-Bromoisophthalic acid ≥98.0% (by GC, titration analysis)
5-Bromoisophthalic acid
Formula: C₈H₅BrO₄
MW: 245.03 g/mol
MDL Number: MFCD00152019
CAS Number: 23351-91-9
EINECS: 245-602-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR