2,6-Diisopropylphenyl isothiocyanate ≥95.0% (by GC)

Supplier: TCI America
Danger

Synonyms: 1,3-Diisopropyl-2-isothiocyanatobenzene

D5830-1G D5830-5G
TCD5830-1GEA 26 USD
TCD5830-1G TCD5830-5G
2,6-Diisopropylphenyl isothiocyanate ≥95.0% (by GC)
2,6-Diisopropylphenyl isothiocyanate
Formula: C₁₃H₁₇NS
MW: 219.35 g/mol
Boiling Pt: 145°C
MDL Number: MFCD00041341
CAS Number: 25343-70-8
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR