(2,6-Difluorophenyl)acetonitrile ≥98.0% (by GC)

Supplier: TCI America
Danger

D4510-25G D4510-5G
TCD4510-25GEA 418 USD
TCD4510-25G TCD4510-5G
(2,6-Difluorophenyl)acetonitrile ≥98.0% (by GC)
(2,6-Difluorophenyl)acetonitrile
Formula: C₈H₅F₂N
Boiling Pt: 109°C
Flash Pt: 53°C
MDL Number: MFCD00009973
CAS Number: 654-01-3
EINECS: 211-504-7
UN: 1992
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR