2,4-Dimethoxyphenyl isothiocyanate ≥98.0% (by GC)

Supplier: TCI America
Danger

Synonyms: 1,3-Dimethoxy-4-isothiocyanatobenzene, 2,4-Dimethoxyphenylisothiocyanate

D4943-25G D4943-5G
TCD4943-25GEA 342 USD
TCD4943-25G TCD4943-5G
2,4-Dimethoxyphenyl isothiocyanate ≥98.0% (by GC)
2,4-Dimethoxyphenyl isothiocyanate
Formula: C₉H₉NO₂S
MW: 195.24 g/mol
MDL Number: MFCD00041058
CAS Number: 33904-03-9
UN: 1759
ADR: 8,III

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 52.0 to 56.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR