2,4,5-Trifluorobenzenesulfonyl chloride ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

Synonyms: 2,4,5-Trifluorobenzene-1-sulphonyl chloride

T3128-1G T3128-5G
TCT3128-1GEA 62 USD
TCT3128-1G TCT3128-5G
2,4,5-Trifluorobenzenesulfonyl chloride ≥98.0% (by GC, titration analysis)
2,4,5-Trifluorobenzenesulphonyl chloride
Formula: C₆H₂ClF₃O₂S
MW: 230.59 g/mol
Flash Pt: 100°C
MDL Number: MFCD00077600
CAS Number: 220227-21-4
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR