2,6-Dichlorocinnamic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 3-(2,6-Dichlorophenyl)acrylic acid, (2E)-3-(2,6-Dichlorophenyl)acrylic acid

D4048-25G D4048-5G
TCD4048-25GEA 106 USD
TCD4048-25G TCD4048-5G
2,6-Dichlorocinnamic acid ≥98.0% (by GC, titration analysis)
2,6-Dichlorocinnamic acid
Formula: C₉H₆Cl₂O₂
MW: 217.05 g/mol
MDL Number: MFCD00004374
CAS Number: 5345-89-1
EINECS: 226-301-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 194.0 to 198.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR