Pentafluorophenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

P1904-1G P1904-5G
TCP1904-1GEA 42 USD
TCP1904-1G TCP1904-5G
Pentafluorophenylboronic acid (contains varying amounts of Anhydride)
Pentafluorophenylboronic acid
Formula: C₆H₂BF₅O₂
Melting Pt: 290°C
MDL Number: MFCD01074663
CAS Number: 1582-24-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 110.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR