4-Chloro-3-nitrobenzamide ≥98.0% (by GC)

Supplier: TCI America
Warning

C3176-1G C3176-5G
TCC3176-1GEA 62 USD
TCC3176-1G TCC3176-5G
4-Chloro-3-nitrobenzamide ≥98.0% (by GC)
4-Chloro-3-nitrobenzamide
Formula: C₇H₅ClN₂O₃
MDL Number: MFCD00017134
CAS Number: 16588-06-0
EINECS: 240-644-1

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 152.0 to 156.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR