4-Bromo-2-fluoropyridine ≥98.0% (by GC)

Supplier: TCI America
Warning

B4778-1G B4778-5G
TCB4778-1GEA 88.9 USD
TCB4778-1G TCB4778-5G
4-Bromo-2-fluoropyridine ≥98.0% (by GC)
4-Bromo-2-fluoropyridine
Formula: C₅H₃BrFN
MW: 175.99 g/mol
Boiling Pt: 65°C
Flash Pt: 71°C
MDL Number: MFCD04112504
CAS Number: 128071-98-7

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR