5-Methyl-2-phenylpyridine ≥98.0% (by GC)
Supplier: TCI America
M2969-1G
M2969-200MG
TCM2969-1GEA
134
USD
TCM2969-1G
TCM2969-200MG
5-Methyl-2-phenylpyridine ≥98.0% (by GC)
5-Methyl-2-phenylpyridine
Formula:
C₁₂H₁₁N |
MDL Number:
MFCD04114180 CAS Number: 27012-22-2 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 53.0 to 57.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...