3-Bromo-2-nitropyridine ≥98.0% (by GC)

Supplier: TCI America
Warning

B4690-1G B4690-5G
TCB4690-1GEA 50 USD
TCB4690-1G TCB4690-5G
3-Bromo-2-nitropyridine ≥98.0% (by GC)
3-Bromo-2-nitropyridine
Formula: C₅H₃BrN₂O₂
MW: 203 g/mol
MDL Number: MFCD00955614
CAS Number: 54231-33-3

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 102.0 to 106.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR