3-Bromo-2,5-dichloropyridine ≥98.0% (by GC)

Supplier: TCI America
Warning

B4832-1G B4832-5G
TCB4832-1GEA 105 USD
TCB4832-1G TCB4832-5G
3-Bromo-2,5-dichloropyridine ≥98.0% (by GC)
3-Bromo-2,5-dichloropyridine
Formula: C₅H₂BrCl₂N
Storage Temperature: Refrigerator
MDL Number: MFCD06659484
CAS Number: 138006-41-4

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 37.0 to 41.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR