5-Amino-2,3-dichloropyridine ≥98.0% (by GC, total nitrogen)

Supplier: TCI America
Warning

A2824-1G A2824-5G
TCA2824-1GEA 73 USD
TCA2824-1G TCA2824-5G
5-Amino-2,3-dichloropyridine ≥98.0% (by GC, total nitrogen)
5-Amino-2,3-dichloropyridine
Formula: C₅H₄Cl₂N₂
Storage Temperature: Refrigerator
MDL Number: MFCD03840434
CAS Number: 98121-41-6

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (with Total Nitrogen) min. 98.0 %
Melting point 111.0 to 115.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR