6-Methoxyanthranilic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Warning
|
A2847-1G
A2847-5G
TCA2847-1GEA
398
USD
TCA2847-1G
TCA2847-5G
6-Methoxyanthranilic acid ≥98.0% (by HPLC, titration analysis)
6-Methoxyanthranilic acid
Formula:
C₈H₉NO₃ MW: 167.16 g/mol |
MDL Number:
MFCD01941328 CAS Number: 53600-33-2 |
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 86.0 to 90.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...