3-Bromo-α,α,α-trifluoro-5-nitrotoluene ≥98.0% (by GC)

Supplier: TCI America

B5333-25G B5333-5G
TCB5333-25GEA 71 USD
TCB5333-25G TCB5333-5G
3-Bromo-α,α,α-trifluoro-5-nitrotoluene ≥98.0% (by GC)
3-Bromo-α,α,α-trifluoro-5-nitrotoluene
Formula: C₇H₃BrF₃NO₂
MDL Number: MFCD03788719
CAS Number: 630125-49-4

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR