2-Bromo-α,α,α,6-tetrafluorotoluene ≥97.0% (by GC)
Supplier: TCI America
|
Warning
|
Synonyms:
1-Bromo-3-fluoro-2-(trifluoromethyl)benzene, 2-Bromo-6-fluorobenzotrifluoride
B4833-1G
B4833-5G
TCB4833-1GEA
30
USD
TCB4833-1G
TCB4833-5G
2-Bromo-α,α,α,6-tetrafluorotoluene ≥97.0% (by GC)
2-Bromo-α,α,α,6-tetrafluorotoluene
Formula:
C₇H₃BrF₄ MW: 243 g/mol |
MDL Number:
MFCD01631569 CAS Number: 261951-85-3 |
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...