3-Bromo-4-iodotoluene ≥98.0% (by GC)

Supplier: TCI America
Warning

B5284-25G B5284-5G
TCB5284-25GEA 73 USD
TCB5284-25G TCB5284-5G
3-Bromo-4-iodotoluene ≥98.0% (by GC)
3-Bromo-4-iodotoluene
Formula: C₇H₆BrI
Boiling Pt: 267°C
MDL Number: MFCD00079718
CAS Number: 71838-16-9

Order Now

Specification Test Results

Appearance Light yellow to Brown clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR