p-Terphenyl ≥99.5% (by GC), purified by sublimation

Supplier: TCI America
Warning

Synonyms: 4-Phenyl-1,1'-biphenyl, 1,4-Diphenylbenzene, 4-Phenylbiphenyl, 1,4-Terphenyl

T3263-25G T3263-5G
TCT3263-25GEA 210.3 USD
TCT3263-25G TCT3263-5G
p-Terphenyl ≥99.5% (by GC), purified by sublimation
p-Terphenyl
Formula: C₁₈H₁₄
MW: 230.31 g/mol
Boiling Pt: 389°C
MDL Number: MFCD00003061
CAS Number: 92-94-4
EINECS: 202-205-2

Order Now

Specification Test Results

Appearance White powder to crystal
Purity (GC) min. 99.5 %
Melting point 212.0 to 216.0 deg-C
Solubility in hot Toluene almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR