3,5-Xylyl isothiocyanate ≥97.0% (by GC)
Supplier: TCI America
|
Danger
|
Synonyms:
3,5-Dimethylphenyl isothiocyanate, 3,5-Dimethylphenylisothiocyanate
D4944-25G
D4944-5G
TCD4944-25GEA
232.2
USD
TCD4944-25G
TCD4944-5G
3,5-Xylyl isothiocyanate ≥97.0% (by GC)
3,5-Xylyl isothiocyanate
Formula:
C₉H₉NS MW: 163.24 g/mol |
MDL Number:
MFCD00041082 CAS Number: 40046-30-8 UN: 2810 ADR: 6.1,III |
Specification Test Results
Appearance | Light orange to Yellow to Green clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...