(2-Ethoxyphenyl)acetic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
E1234-1G
E1234-5G
TCE1234-1GEA
38
USD
TCE1234-1G
TCE1234-5G
(2-Ethoxyphenyl)acetic acid ≥98.0% (by GC, titration analysis)
(2-Ethoxyphenyl)acetic acid
Formula:
H₃CCH₂O(C₆H₄)CH₂COOH |
MDL Number:
MFCD00016824 CAS Number: 70289-12-2 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 103.0 to 107.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...