1,3-Benzenedithiol ≥95.0% (by GC)
Supplier: TCI America
|
Warning
|
B4371-1G
B4371-5G
TCB4371-1GEA
147.5
USD
TCB4371-1G
TCB4371-5G
1,3-Benzenedithiol ≥95.0% (by GC)
1,3-Benzenedithiol
Formula:
C₆H₆S₂ Boiling Pt: 245°C Melting Pt: 27°C |
MDL Number:
MFCD00004842 CAS Number: 626-04-0 EINECS: 210-925-3 |
Specification Test Results
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...