Dimethylammonium bromide ≥98.0% (by total nitrogen and titration analysis)

Supplier: TCI America
Warning

D5092-1G D5092-5G
TCD5092-1GEA 44 USD
TCD5092-1G TCD5092-5G
Dimethylammonium bromide ≥98.0% (by total nitrogen and titration analysis)
Dimethylammonium bromide
Formula: C₂H₈BrN
MDL Number: MFCD00051983
CAS Number: 6912-12-5
EINECS: 230-019-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR