2-Fluoro-5-hydroxypyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 2-Fluoro-5-pyridinol, 6-Fluoropyridin-3-ol

F0980-1G F0980-5G
TCF0980-1GEA 74.3 USD
TCF0980-1G TCF0980-5G
2-Fluoro-5-hydroxypyridine ≥98.0% (by GC, titration analysis)
2-Fluoro-5-hydroxypyridine
Formula: C₅H₄FNO
MDL Number: MFCD01075040
CAS Number: 55758-32-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 151.0 to 155.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR