1-Methylisatin ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 1-Methylindoline-2,3-dione, 1-Methyl-2,3-dihydroindole-2,3-dione, N-Methylindoline-2,3-dione, N-Methylisatin, 1-Methyl-1H-indole-2,3-dione, N-Methyl-1H-indole-2,3-dione, N-Methyl-2,3-dihydroindole-2,3-dione

M2706-1G M2706-5G
TCM2706-1GEA 29 USD
TCM2706-1G TCM2706-5G
1-Methylisatin ≥98.0% (by GC)
1-Methylisatin
Formula: C₉H₇NO₂
MW: 161.16 g/mol
MDL Number: MFCD00005812
CAS Number: 2058-74-4
EINECS: 218-164-9

Order Now

Specification Test Results

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 128.0 to 133.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR