2-Amino-4-chloropyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

A2152-25G A2152-5G
TCA2152-25GEA 214 USD
TCA2152-25G TCA2152-5G
2-Amino-4-chloropyridine ≥98.0% (by GC, titration analysis)
2-Amino-4-chloropyridine
Formula: C₅H₅ClN₂
MDL Number: MFCD04113820
CAS Number: 19798-80-2

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 130.0 to 134.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR