3-Chloro-2-iodotoluene ≥95.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 1-Chloro-2-iodo-3-methylbenzene

C3364-25G C3364-5G
TCC3364-25GEA 184 USD
TCC3364-25G TCC3364-5G
3-Chloro-2-iodotoluene ≥95.0% (by GC)
3-Chloro-2-iodotoluene
Formula: C₇H₆ClI
MW: 252.48 g/mol
Boiling Pt: 125°C
Storage Temperature: Refrigerator
MDL Number: MFCD00051756
CAS Number: 5100-98-1

Order Now

Specification Test Results

Appearance White to Light yellow to Dark green powder to lump
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR