(2,4-Dichlorophenyl)acetic acid methyl ester ≥99.0% (by GC)

Supplier: TCI America

Synonyms: Methyl 2,4-Dichlorophenylacetate, Methyl (2,4-dichlorophenyl)acetate, Methyl 2,4-dichlorophenylacetate, 2,4-Dichlorophenylacetic acid methyll ester

M2528-25G M2528-5G
TCM2528-25GEA 125.5 USD
TCM2528-25G TCM2528-5G
(2,4-Dichlorophenyl)acetic acid methyl ester ≥99.0% (by GC)
(2,4-Dichlorophenyl)acetic acid methyl ester
Formula: (C₆H₃Cl₂)CH₂COOCH₃
MW: 219.07 g/mol
Boiling Pt: 153°C
MDL Number: MFCD00018381
CAS Number: 55954-23-9

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR