5,6-Dimethoxyindole ≥98.0% (by GC)

Supplier: TCI America
Warning

D5148-1G D5148-200MG
TCD5148-1GEA 266 USD
TCD5148-1G TCD5148-200MG
5,6-Dimethoxyindole ≥98.0% (by GC)
5,6-Dimethoxyindole
Formula: C₁₀H₁₁NO₂
MW: 177.2 g/mol
Boiling Pt: 198°C
MDL Number: MFCD00005675
CAS Number: 14430-23-0
EINECS: 238-402-5

Order Now

Specification Test Results

Appearance White to Light gray to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 153.0 to 157.0 deg-C
Solubility in hot Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR