2-(Trifluoromethoxy)benzylamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

Synonyms: (2-(Trifluoromethoxy)phenyl)methanamine

T3092-1G T3092-5G
TCT3092-1GEA 42 USD
TCT3092-1G TCT3092-5G
2-(Trifluoromethoxy)benzylamine ≥98.0% (by GC, titration analysis)
2-(Trifluoromethoxy)benzylamine
Formula: C₈H₈F₃NO
MDL Number: MFCD00085176
CAS Number: 175205-64-8
UN: 2735
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid to cloudy liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR