2-(Trifluoromethyl)benzamide ≥98.0% (by GC)

Supplier: TCI America

T3384-25G T3384-5G
76220-600EA 268 USD
76220-600 76220-602
2-(Trifluoromethyl)benzamide ≥98.0% (by GC)
2-(Trifluoromethyl)benzamide
Formula: C₈H₆F₃NO
MDL Number: MFCD00014798
CAS Number: 360-64-5
EINECS: 206-637-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 162.0 to 166.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR