1-Phenylisatin ≥98.0% (by GC)

Supplier: TCI America
Warning

P2279-25G P2279-5G
TCP2279-25GEA 574 USD
TCP2279-25G TCP2279-5G
1-Phenylisatin ≥98.0% (by GC)
1-Phenylisatin
Formula: C₁₄H₉NO₂
MDL Number: MFCD00082681
CAS Number: 723-89-7

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 139.0 to 143.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR