2-Amino-5-bromo-3-iodopyridine ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

A2563-1G A2563-5G
TCA2563-1GEA 69 USD
TCA2563-1G TCA2563-5G
2-Amino-5-bromo-3-iodopyridine ≥98.0% (by titrimetric analysis)
2-Amino-5-bromo-3-iodopyridine
Formula: C₅H₄BrIN₂
MDL Number: MFCD06659000
CAS Number: 381233-96-1

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 96.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point gt. 112.0 and le 116.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR