2-Bromo-4-iodopyridine ≥98.0% (by GC)

Supplier: TCI America
Warning

B5119-1G B5119-5G
TCB5119-1GEA 74 USD
TCB5119-1G TCB5119-5G
2-Bromo-4-iodopyridine ≥98.0% (by GC)
2-Bromo-4-iodopyridine
Formula: C₅H₃BrIN
MDL Number: MFCD03085768
CAS Number: 100523-96-4

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 62.0 to 66.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR