2-Methyl-6-quinolinol ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 6-Hydroxy-2-methylquinoline

H1446-1G H1446-5G
TCH1446-1GEA 104 USD
TCH1446-1G TCH1446-5G
2-Methyl-6-quinolinol ≥98.0% (by GC, titration analysis)
2-Methyl-6-quinolinol
Formula: C₁₀H₉NO
MW: 159.19 g/mol
MDL Number: MFCD00047600
CAS Number: 613-21-8

Order Now

Specification Test Results

Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 214.0 to 218.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR