4-Chloroquinoline ≥97.0% (by GC)
Supplier: TCI America
|
Warning
|
C2284-1G
C2284-5G
TCC2284-1GEA
58
USD
TCC2284-1G
TCC2284-5G
4-Chloroquinoline ≥97.0% (by GC)
4-Chloroquinoline
Formula:
C₉H₆ClN Boiling Pt: 261°C Storage Temperature: Refrigerator |
MDL Number:
MFCD00006773 CAS Number: 611-35-8 EINECS: 210-267-7 |
Specification Test Results
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...