2-Quinolinecarbonitrile ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: Quinoline-2-carbonitrile

Q0099-1G Q0099-5G
TCQ0099-1GEA 71 USD
TCQ0099-1G TCQ0099-5G
2-Quinolinecarbonitrile ≥98.0% (by GC, titration analysis)
2-Quinolinecarbonitrile
Formula: C₁₀H₆N₂
MW: 154.17 g/mol
Boiling Pt: 160°C
MDL Number: MFCD00134341
CAS Number: 1436-43-7
EINECS: 215-865-1

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 93.0 to 97.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR