2,6-Dimethylbenzonitrile ≥98.0% (by GC)

Supplier: TCI America
Warning

D4927-25G D4927-5G
76219-758EA 285 USD
76219-758 76219-760
2,6-Dimethylbenzonitrile ≥98.0% (by GC)
2,6-Dimethylbenzonitrile
Formula: C₉H₉N
MW: 131.18 g/mol
Boiling Pt: 93°C
MDL Number: MFCD00037166
CAS Number: 6575-13-9
EINECS: 229-503-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 88.0 to 92.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR