(2-Fluorophenyl)acetone ≥97.0% (by GC)

Supplier: TCI America

Synonyms: 2-Fluorophenylacetone, 1-(2-Fluorophenyl)propan-2-one, 1-(2-Fluorophenyl)-2-propanone

F0970-25G F0970-5G
TCF0970-25GEA 233.3 USD
TCF0970-25G TCF0970-5G
(2-Fluorophenyl)acetone ≥97.0% (by GC)
(2-Fluorophenyl)acetone
Formula: C₉H₉FO
MW: 152.17 g/mol
Boiling Pt: 47°C
Flash Pt: 85°C
MDL Number: MFCD00000325
CAS Number: 2836-82-0
EINECS: 220-627-5

Order Now

Specification Test Results

Appearance White to Yellow to Green clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR