3-Hydroxy-4-iodobenzoic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

H1496-1G H1496-5G
TCH1496-1GEA 110 USD
TCH1496-1G TCH1496-5G
3-Hydroxy-4-iodobenzoic acid ≥98.0% (by HPLC, titration analysis)
3-Hydroxy-4-iodobenzoic acid
Formula: C₇H₅IO₃
MW: 264.02 g/mol
MDL Number: MFCD02068386
CAS Number: 58123-77-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 225.0 to 233.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR