4-Hydroxy-3,5-diiodobenzaldehyde ≥97.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
3,5-Diiodo4-hydroxybenzaldehyde, 3,5-Diiodo-4-hydroxybenzaldehyde
H1539-25G
H1539-5G
TCH1539-25GEA
183
USD
TCH1539-25G
TCH1539-5G
4-Hydroxy-3,5-diiodobenzaldehyde ≥97.0% (by titrimetric analysis)
4-Hydroxy-3,5-diiodobenzaldehyde
Formula:
C₇H₄I₂O₂ MW: 373.92 g/mol |
MDL Number:
MFCD00014670 CAS Number: 1948-40-9 EINECS: 217-754-3 |
Specification Test Results
Purity (GC) | min. 96.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Melting point | 202.0 to 206.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...